Anti-PGAM2 Mouse Monoclonal Antibody [clone: 2F5]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF503424EA 5590 DKK
ORIGCF503424
Anti-PGAM2 Mouse Monoclonal Antibody [clone: 2F5]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:50-100

Type: Primary
Antigen: PGAM2
Clonality: Monoclonal
Clone: 2F5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR