Anti-CD1C Mouse Monoclonal Antibody [clone: 1H9]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF505384EA 5590 DKK
ORIGCF505384
Anti-CD1C Mouse Monoclonal Antibody [clone: 1H9]
Antistoffer
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: CD1C
Clonality: Monoclonal
Clone: 1H9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human