Anti-MAGEA4 Mouse Monoclonal Antibody [clone: 2C1]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF505361EA 5590 DKK
ORIGCF505361
Anti-MAGEA4 Mouse Monoclonal Antibody [clone: 2C1]
Antistoffer
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: MAGEA4
Clonality: Monoclonal
Clone: 2C1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human