Anti-KCNJ3 Mouse Monoclonal Antibody [clone: 1B1]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504193EA 5590 DKK
ORIGCF504193
Anti-KCNJ3 Mouse Monoclonal Antibody [clone: 1B1]
Antistoffer
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: KCNJ3
Clonality: Monoclonal
Clone: 1B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR