Anti-DOK2 Mouse Monoclonal Antibody [clone: 2D10]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF503381EA 5590 DKK
ORIGCF503381
Anti-DOK2 Mouse Monoclonal Antibody [clone: 2D10]
Antistoffer
Recommended Dilutions: Western Blot: 1:200-500

Type: Primary
Antigen: DOK2
Clonality: Monoclonal
Clone: 2D10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR