Anti-CCT8L2 Mouse Monoclonal Antibody [clone: 1A8]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF505336EA 5750 DKK
ORIGCF505336
Anti-CCT8L2 Mouse Monoclonal Antibody [clone: 1A8]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100
Type: Primary
Antigen: CCT8L2
Clonality: Monoclonal
Clone: 1A8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human