Anti-RPS6KB1 Mouse Monoclonal Antibody [clone: 6B2]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF502929EA 5590 DKK
ORIGCF502929
Anti-RPS6KB1 Mouse Monoclonal Antibody [clone: 6B2]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: RPS6KB1
Clonality: Monoclonal
Clone: 6B2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse