Anti-KCNJ3 Mouse Monoclonal Antibody [clone: 1C5]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504138EA 5590 DKK
ORIGCF504138
Anti-KCNJ3 Mouse Monoclonal Antibody [clone: 1C5]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: KCNJ3
Clonality: Monoclonal
Clone: 1C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR