Anti-USP10 Mouse Monoclonal Antibody [clone: 1A10]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504130EA 5750 DKK
ORIGCF504130
Anti-USP10 Mouse Monoclonal Antibody [clone: 1A10]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: USP10
Clonality: Monoclonal
Clone: 1A10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR