Anti-RNF113B Mouse Monoclonal Antibody [clone: 2A6]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504119EA 5590 DKK
ORIGCF504119
Anti-RNF113B Mouse Monoclonal Antibody [clone: 2A6]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: RNF113B
Clonality: Monoclonal
Clone: 2A6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR