Anti-PDLIM2 Mouse Monoclonal Antibody [clone: 15C1]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF502874EA 5590 DKK
ORIGCF502874
Anti-PDLIM2 Mouse Monoclonal Antibody [clone: 15C1]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PDLIM2
Clonality: Monoclonal
Clone: 15C1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR