Anti-PSMB4 Mouse Monoclonal Antibody [clone: 2H2]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504090EA 5590 DKK
ORIGCF504090
Anti-PSMB4 Mouse Monoclonal Antibody [clone: 2H2]
Antistoffer
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: PSMB4
Clonality: Monoclonal
Clone: 2H2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR