Anti-USP7 Mouse Monoclonal Antibody [clone: 3D5]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504061EA 5590 DKK
ORIGCF504061
Anti-USP7 Mouse Monoclonal Antibody [clone: 3D5]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: USP7
Clonality: Monoclonal
Clone: 3D5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR