Anti-PARVA Mouse Monoclonal Antibody [clone: 3C3]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF505992EA 5590 DKK
ORIGCF505992
Anti-PARVA Mouse Monoclonal Antibody [clone: 3C3]
Antistoffer
Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150
Type: Primary
Antigen: PARVA
Clonality: Monoclonal
Clone: 3C3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse