Anti-UBE2G2 Mouse Monoclonal Antibody [clone: 5F1]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505239EA 5750 DKK
ORIGCF505239
Anti-UBE2G2 Mouse Monoclonal Antibody [clone: 5F1]
Antistoffer
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: UBE2G2
Clonality: Monoclonal
Clone: 5F1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR