Anti-FATE1 Mouse Monoclonal Antibody [clone: 6A11]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505216EA 5590 DKK
ORIGCF505216
Anti-FATE1 Mouse Monoclonal Antibody [clone: 6A11]
Antistoffer
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: FATE1
Clonality: Monoclonal
Clone: 6A11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR