Anti-FATE1 Mouse Monoclonal Antibody [clone: 6F10]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF505215EA 5590 DKK
ORIGCF505215
Anti-FATE1 Mouse Monoclonal Antibody [clone: 6F10]
Antistoffer
Recommended Dilutions: Western Blot: 1:200-4000; Immunofluorescence: 1:100
Type: Primary
Antigen: FATE1
Clonality: Monoclonal
Clone: 6F10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat