Anti-DOK7 Mouse Monoclonal Antibody [clone: 3C12]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504832EA 5590 DKK
ORIGCF504832
Anti-DOK7 Mouse Monoclonal Antibody [clone: 3C12]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: DOK7
Clonality: Monoclonal
Clone: 3C12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR