Anti-FBXO21 Mouse Monoclonal Antibody [clone: 5H4]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504025EA 5750 DKK
ORIGCF504025
Anti-FBXO21 Mouse Monoclonal Antibody [clone: 5H4]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: FBXO21
Clonality: Monoclonal
Clone: 5H4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR