Anti-NLN Mouse Monoclonal Antibody [clone: 3C2]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504021EA 5590 DKK
ORIGCF504021
Anti-NLN Mouse Monoclonal Antibody [clone: 3C2]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: NLN
Clonality: Monoclonal
Clone: 3C2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR