Anti-RASSF8 Mouse Monoclonal Antibody [clone: 3F7]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF505906EA 5590 DKK
ORIGCF505906
Anti-RASSF8 Mouse Monoclonal Antibody [clone: 3F7]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: RASSF8
Clonality: Monoclonal
Clone: 3F7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey