Anti-DPCD Mouse Monoclonal Antibody [clone: 4B9]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505898EA 5750 DKK
ORIGCF505898
Anti-DPCD Mouse Monoclonal Antibody [clone: 4B9]
Antistoffer
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: DPCD
Clonality: Monoclonal
Clone: 4B9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR