Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 2G3]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF505890EA 5590 DKK
ORIGCF505890
Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 2G3]
Antistoffer
Recommended Dilutions: Western Blot: 1:200-2000; Immunofluorescence: 1:100
Type: Primary
Antigen: MAP2K3
Clonality: Monoclonal
Clone: 2G3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey