Anti-ERBB2 Mouse Monoclonal Antibody [clone: 7C2]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505883EA 5590 DKK
ORIGCF505883
Anti-ERBB2 Mouse Monoclonal Antibody [clone: 7C2]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: 7C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR