Anti-NDUFB10 Mouse Monoclonal Antibody [clone: 2H10]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505171EA 5750 DKK
ORIGCF505171
Anti-NDUFB10 Mouse Monoclonal Antibody [clone: 2H10]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: NDUFB10
Clonality: Monoclonal
Clone: 2H10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR