Anti-DOK7 Mouse Monoclonal Antibody [clone: 5C6]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504788EA 5590 DKK
ORIGCF504788
Anti-DOK7 Mouse Monoclonal Antibody [clone: 5C6]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150

Type: Primary
Antigen: DOK7
Clonality: Monoclonal
Clone: 5C6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR