Anti-CALR3 Mouse Monoclonal Antibody [clone: 1D2]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505837EA 5590 DKK
ORIGCF505837
Anti-CALR3 Mouse Monoclonal Antibody [clone: 1D2]
Antistoffer
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: CALR3
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR