Anti-HGS Mouse Monoclonal Antibody [clone: 1C2]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505832EA 5750 DKK
ORIGCF505832
Anti-HGS Mouse Monoclonal Antibody [clone: 1C2]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: HGS
Clonality: Monoclonal
Clone: 1C2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR