Anti-SRPRB Mouse Monoclonal Antibody [clone: 3E6]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF504715EA 5750 DKK
ORIGCF504715
Anti-SRPRB Mouse Monoclonal Antibody [clone: 3E6]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500
Type: Primary
Antigen: SRPRB
Clonality: Monoclonal
Clone: 3E6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human