Anti-UBE2E3 Mouse Monoclonal Antibody [clone: 4G4]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504683EA 5590 DKK
ORIGCF504683
Anti-UBE2E3 Mouse Monoclonal Antibody [clone: 4G4]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: UBE2E3
Clonality: Monoclonal
Clone: 4G4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR