Anti-METT10D Mouse Monoclonal Antibody [clone: 3E8]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504665EA 5750 DKK
ORIGCF504665
Anti-METT10D Mouse Monoclonal Antibody [clone: 3E8]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500

Type: Primary
Antigen: METT10D
Clonality: Monoclonal
Clone: 3E8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR