Anti-ADIPOQ Mouse Monoclonal Antibody [clone: 7B11]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF503913EA 5590 DKK
ORIGCF503913
Anti-ADIPOQ Mouse Monoclonal Antibody [clone: 7B11]
Antistoffer
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100; Western Blot: 1:200

Type: Primary
Antigen: ADIPOQ
Clonality: Monoclonal
Clone: 7B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR