Anti-ESRRG Mouse Monoclonal Antibody [clone: 5G8]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505036EA 5590 DKK
ORIGCF505036
Anti-ESRRG Mouse Monoclonal Antibody [clone: 5G8]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: ESRRG
Clonality: Monoclonal
Clone: 5G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR