Anti-UBE2J1 Mouse Monoclonal Antibody [clone: 2G2]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505035EA 5590 DKK
ORIGCF505035
Anti-UBE2J1 Mouse Monoclonal Antibody [clone: 2G2]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: UBE2J1
Clonality: Monoclonal
Clone: 2G2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR