Anti-TMEM173 Mouse Monoclonal Antibody [clone: 4A7]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505022EA 5590 DKK
ORIGCF505022
Anti-TMEM173 Mouse Monoclonal Antibody [clone: 4A7]
Antistoffer
Recommended Dilutions: Western Blot: 1:200-1:1000

Type: Primary
Antigen: TMEM173
Clonality: Monoclonal
Clone: 4A7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR