Anti-APP Mouse Monoclonal Antibody [clone: 7A10]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505012EA 5590 DKK
ORIGCF505012
Anti-APP Mouse Monoclonal Antibody [clone: 7A10]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150

Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone: 7A10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR