Anti-C17orf37 Mouse Monoclonal Antibody [clone: 4D6]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504625EA 5590 DKK
ORIGCF504625
Anti-C17orf37 Mouse Monoclonal Antibody [clone: 4D6]
Antistoffer
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: C17orf37
Clonality: Monoclonal
Clone: 4D6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR