Anti-LDLRAP1 Mouse Monoclonal Antibody [clone: 7G2]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF503841EA 5590 DKK
ORIGCF503841
Anti-LDLRAP1 Mouse Monoclonal Antibody [clone: 7G2]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LDLRAP1
Clonality: Monoclonal
Clone: 7G2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR