Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 2A1]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505763EA 5590 DKK
ORIGCF505763
Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 2A1]
Antistoffer
Recommended Dilutions: Western Blot: 1:400-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K1
Clonality: Monoclonal
Clone: 2A1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR