Anti-LRRC50 Mouse Monoclonal Antibody [clone: 4H3]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504578EA 5590 DKK
ORIGCF504578
Anti-LRRC50 Mouse Monoclonal Antibody [clone: 4H3]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: LRRC50
Clonality: Monoclonal
Clone: 4H3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR