Anti-CDC123 Mouse Monoclonal Antibody [clone: 5C7]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF505694EA 5590 DKK
ORIGCF505694
Anti-CDC123 Mouse Monoclonal Antibody [clone: 5C7]
Antistoffer
Recommended Dilutions: Western Blot: 1:400-4000; Immunofluorescence: 1:100
Type: Primary
Antigen: CDC123
Clonality: Monoclonal
Clone: 5C7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse