Anti-RAB24 Mouse Monoclonal Antibody [clone: 3H1]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505690EA 5590 DKK
ORIGCF505690
Anti-RAB24 Mouse Monoclonal Antibody [clone: 3H1]
Antistoffer
Recommended Dilutions: Western Blot: 1:200-4000

Type: Primary
Antigen: RAB24
Clonality: Monoclonal
Clone: 3H1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR