Anti-COMMD1 Mouse Monoclonal Antibody [clone: 2D10]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF504920EA 5590 DKK
ORIGCF504920
Anti-COMMD1 Mouse Monoclonal Antibody [clone: 2D10]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: COMMD1
Clonality: Monoclonal
Clone: 2D10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Rat