Anti-VBP1 Mouse Monoclonal Antibody [clone: 2E6]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504914EA 5590 DKK
ORIGCF504914
Anti-VBP1 Mouse Monoclonal Antibody [clone: 2E6]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: VBP1
Clonality: Monoclonal
Clone: 2E6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR