Anti-VBP1 Mouse Monoclonal Antibody [clone: 2B12]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504904EA 5590 DKK
ORIGCF504904
Anti-VBP1 Mouse Monoclonal Antibody [clone: 2B12]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: VBP1
Clonality: Monoclonal
Clone: 2B12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR