Anti-EIF2B3 Mouse Monoclonal Antibody [clone: 1A4]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF503758EA 5590 DKK
ORIGCF503758
Anti-EIF2B3 Mouse Monoclonal Antibody [clone: 1A4]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: EIF2B3
Clonality: Monoclonal
Clone: 1A4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human