Anti-HRAS Mouse Monoclonal Antibody [clone: 1H3]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505674EA 5590 DKK
ORIGCF505674
Anti-HRAS Mouse Monoclonal Antibody [clone: 1H3]
Antistoffer
Recommended Dilutions: Western Blot: 1:200-4000

Type: Primary
Antigen: HRAS
Clonality: Monoclonal
Clone: 1H3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR