Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 4G4]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505659EA 5590 DKK
ORIGCF505659
Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 4G4]
Antistoffer
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K3
Clonality: Monoclonal
Clone: 4G4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR