Anti-MLF2 Mouse Monoclonal Antibody [clone: 1E12]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504875EA 5750 DKK
ORIGCF504875
Anti-MLF2 Mouse Monoclonal Antibody [clone: 1E12]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: MLF2
Clonality: Monoclonal
Clone: 1E12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR