Anti-S100P Mouse Monoclonal Antibody [clone: 1A4]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF504478EA 5750 DKK
ORIGCF504478
Anti-S100P Mouse Monoclonal Antibody [clone: 1A4]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100
Type: Primary
Antigen: S100P
Clonality: Monoclonal
Clone: 1A4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human